Please login to your online account to display your discounted pricing

Mouse anti-NKX2.2, Clone: SPM564, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479901

  • / Each
 

Description & Specifications

Specifications

Format Purified
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Antigen NKX2.2
Purification Method Purified
Regulatory Status RUO
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM564
Immunogen Human NKX2.2 recombinant protein
Concentration 1mg/mL
Reconstitution Protein A purified
Gene ID (Entrez) 4821
Gene Accession No. O95096
Primary or Secondary Primary
Host Species Mouse
Isotype IgG2b, κ