Please login to your online account to display your discounted pricing

Mouse anti-NF-H, Clone: SPM563, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen NF-H
Clone SPM563
Concentration 0.2mg/mL
Immunogen Human recombinant neurofilament protein
Gene Accession No. P12036
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23438001

 
novus biologicals canada
NBP23438001ML
0.1mL Each for $139.00

NBP23438005

 
novus biologicals canada
NBP23438005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen NF-H
Clone SPM563
Concentration 0.2mg/mL
Immunogen Human recombinant neurofilament protein
Gene Accession No. P12036
Gene ID (Entrez) 4744
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting