Please login to your online account to display your discounted pricing

Mouse anti-NF-H, Clone: SPM563, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438005

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen NF-H
Format Purified
Host Species Mouse
Quantity 0.5mL
Regulatory Status RUO
Test Specificity This MAb reacts with a 200kDa protein, identified as heavy sub-unit of neurofilaments (NF-H).
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P12036
Immunogen Human recombinant neurofilament protein
Isotype IgG1
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 4744
Clone SPM563
Purification Method Purified