Please login to your online account to display your discounted pricing

Mouse anti-NF-H, Clone: SPM563, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438001

  • / Each
 

Description & Specifications

Specifications

Format Purified
Gene Accession No. P12036
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen NF-H
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Immunogen Human recombinant neurofilament protein
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4744
Test Specificity This MAb reacts with a 200kDa protein, identified as heavy sub-unit of neurofilaments (NF-H).
Clone SPM563
Concentration 0.2mg/mL
Conjugate Unlabeled