missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-NF-H, Clone: SPM563, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen NF-H
Clone SPM563
Concentration 1mg/mL
Immunogen Human recombinant neurofilament protein
Gene Accession No. P12036
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23442701 Novus Biologicals Canada
NBP23442701MG
0.1mg Each for $384.00
NBP23442702 Novus Biologicals Canada
NBP23442702MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

NF-H
1mg/mL
P12036
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM563
Human recombinant neurofilament protein
4744
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Purified
ELISA,Flow Cytometry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Western Blot