Please login to your online account to display your discounted pricing

Mouse anti-NF-H, Clone: SPM563, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Human recombinant neurofilament protein
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM563
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Gene Accession No. P12036
Conjugate Unlabeled
Host Species Mouse
Primary or Secondary Primary
Antigen NF-H
Format Purified
Purification Method Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Gene ID (Entrez) 4744
Quantity 0.1mg
Isotype IgG1
Regulatory Status RUO
Test Specificity This MAb reacts with a 200kDa protein, identified as heavy sub-unit of neurofilaments (NF-H).