Please login to your online account to display your discounted pricing

Mouse anti-Neprilysin/CD10, Clone: SN5c/L4-1A1, PE, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen Neprilysin/CD10
Clone SN5c/L4-1A1
Immunogen Native purified MME from membrane preparation of human B-lineage leukemia cells.
Gene ID (Entrez) 4311
Formulation Aqueous buffer (contains stabilizer if necessary)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143891

 
novus biologicals canada
NBP143891100TESTS
100 Tests Each for $252.00

NBP14389125

 
novus biologicals canada
NBP14389125TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen Neprilysin/CD10
Clone SN5c/L4-1A1
Immunogen Native purified MME from membrane preparation of human B-lineage leukemia cells.
Gene ID (Entrez) 4311
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry