Please login to your online account to display your discounted pricing

Mouse anti-Neprilysin/CD10, Clone: SN5c, Biotin, Novus Biologicals™

$96.00 - $192.00

Specifications

Antigen Neprilysin/CD10
Clone SN5c
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD10.
Gene ID (Entrez) 4311
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14368001

 
novus biologicals canada
NBP14368001MG
0.1mg Each for $192.00

NBP14368003

 
novus biologicals canada
NBP1436800025MG
0.025mg Each for $96.00
Description & Specifications

Specifications

Antigen Neprilysin/CD10
Clone SN5c
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD10.
Gene ID (Entrez) 4311
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)