Please login to your online account to display your discounted pricing

Mouse anti-Neprilysin/CD10, Clone: CB-CALLA, Novus Biologicals™

$214.00 - $278.00

Specifications

Antigen Neprilysin/CD10
Clone CB-CALLA
Immunogen The immunogen for this antibody was CD10.
Gene ID (Entrez) 4311
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14344301

 
novus biologicals canada
NBP14344301MG
0.1mg Each for $278.00

NBP14344303

 
novus biologicals canada
NBP1434430025MG
0.025mg Each for $214.00
Description & Specifications

Specifications

Antigen Neprilysin/CD10
Clone CB-CALLA
Immunogen The immunogen for this antibody was CD10.
Gene ID (Entrez) 4311
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry