Please login to your online account to display your discounted pricing

Mouse anti-Neprilysin/CD10, Clone: CB-CALLA, Biotin, Novus Biologicals™

$96.00 - $192.00

Specifications

Antigen Neprilysin/CD10
Clone CB-CALLA
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD10.
Gene ID (Entrez) 4311
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14368101

 
novus biologicals canada
NBP14368101MG
0.1mg Each for $192.00

NBP14368103

 
novus biologicals canada
NBP1436810025MG
0.025mg Each for $96.00
Description & Specifications

Specifications

Antigen Neprilysin/CD10
Clone CB-CALLA
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD10.
Gene ID (Entrez) 4311
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry