Please login to your online account to display your discounted pricing

Mouse anti-Neprilysin/CD10, Clone: CB-CALLA, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1436810025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14368103

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Quantity 0.025mg
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Purification Method Affinity Purified
Applications Flow Cytometry
Cross Reactivity Human
Antigen Neprilysin/CD10
Formulation PBS [pH 7.2]
Isotype IgG2b
Conjugate Biotin
Gene ID (Entrez) 4311
Regulatory Status RUO
Host Species Mouse
Immunogen The immunogen for this antibody was CD10.
Clone CB-CALLA