Please login to your online account to display your discounted pricing

Mouse anti-Neprilysin/CD10, Clone: CB-CALLA, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14368101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14368101

  • / Each
 

Description & Specifications

Specifications

Antigen Neprilysin/CD10
Applications Flow Cytometry
Clone CB-CALLA
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Human
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Mouse
Immunogen The immunogen for this antibody was CD10.
Isotype IgG2b
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4311