Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: SPM489, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen NCAM-1/CD56
Clone SPM489
Concentration 0.2mg/mL
Immunogen Membrane preparation of a small cell lung carcinoma
Gene ID (Entrez) 4684
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23436401

 
novus biologicals canada
NBP23436401ML
0.1mL Each for $139.00

NBP23436405

 
novus biologicals canada
NBP23436405ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen NCAM-1/CD56
Clone SPM489
Concentration 0.2mg/mL
Immunogen Membrane preparation of a small cell lung carcinoma
Gene ID (Entrez) 4684
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting