Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: SPM489, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen NCAM-1/CD56
Clone SPM489
Concentration 1mg/mL
Immunogen Membrane preparation of a small cell lung carcinoma
Gene ID (Entrez) 4684
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23441201

 
novus biologicals canada
NBP23441201MG
0.1mg Each for $384.00

NBP23441202

 
novus biologicals canada
NBP23441202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen NCAM-1/CD56
Clone SPM489
Concentration 1mg/mL
Immunogen Membrane preparation of a small cell lung carcinoma
Gene ID (Entrez) 4684
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting