Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: SPM489, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441202

  • / Each
 

Description & Specifications

Specifications

Quantity 0.2mg
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen NCAM-1/CD56
Immunogen Membrane preparation of a small cell lung carcinoma
Conjugate Unlabeled
Regulatory Status RUO
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Cross Reactivity Human
Gene ID (Entrez) 4684
Clone SPM489
Reconstitution Protein A purified
Host Species Mouse
Concentration 1mg/mL
Test Specificity This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135kDa isoform is the basic molecule, which is glycosylated or sialylated to produce the mature species. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas). It is also expressed on some mesodermally derived tumors (rhabdomyosarcoma). Anti-CD56 plays an important role in the diagnosis of nodal and nasal NK/T-cell lymphomas.
Format Purified
Isotype IgG1, κ
Monoclonal or Polyclonal Monoclonal
Purification Method Purified
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.