Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: SPM128, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434901

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Antigen NCAM-1/CD56
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas).
Immunogen Membrane preparation of a small cell lung carcinoma
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 4684
Reconstitution Protein A purified
Clone SPM128
Concentration 0.2mg/mL