Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: SPM128, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23434905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23434905

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Immunogen Membrane preparation of a small cell lung carcinoma
Format Purified
Isotype IgG1, κ
Regulatory Status RUO
Conjugate Unlabeled
Quantity 0.5mL
Gene ID (Entrez) 4684
Antigen NCAM-1/CD56
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Host Species Mouse
Reconstitution Protein A purified
Purification Method Purified
Clone SPM128
Concentration 0.2mg/mL
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas).