missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: SPM128, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen NCAM-1/CD56
Clone SPM128
Concentration 1mg/mL
Immunogen Membrane preparation of a small cell lung carcinoma
Gene ID (Entrez) 4684
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23439701 Novus Biologicals Canada
NBP23439701MG
0.1mg Each for $384.00
NBP23439702 Novus Biologicals Canada
NBP23439702MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

NCAM-1/CD56
1mg/mL
4684
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
SPM128
Membrane preparation of a small cell lung carcinoma
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified