Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: SPM128, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439702MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439702

  • / Each
 

Description & Specifications

Specifications

Antigen NCAM-1/CD56
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM128
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen Membrane preparation of a small cell lung carcinoma
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4684
Test Specificity This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas).
Reconstitution Protein A purified
Target Species Human
Target Species Rat
Target Species Zebrafish