Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: SPM128, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439701

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas).
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Zebrafish
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4684
Clone SPM128
Concentration 1mg/mL
Conjugate Unlabeled
Antigen NCAM-1/CD56
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Membrane preparation of a small cell lung carcinoma
Quantity 0.1mg
Reconstitution Protein A purified