Please login to your online account to display your discounted pricing

Mouse anti-NCAM-1/CD56, Clone: 123c3.D5+123A8, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen NCAM-1/CD56
Clone 123c3.D5+123A8
Concentration 1mg/mL
Immunogen Membrane preparation of a small cell lung carcinoma
Gene Accession No. P13591
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23451902

 
novus biologicals canada
NBP23451902MG
0.2mg Each for $527.00

NBP23451901

 
novus biologicals canada
NBP23451901MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen NCAM-1/CD56
Clone 123c3.D5+123A8
Concentration 1mg/mL
Immunogen Membrane preparation of a small cell lung carcinoma
Gene Accession No. P13591
Gene ID (Entrez) 4684
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting