Please login to your online account to display your discounted pricing

Mouse anti-Nanog, Clone: hNanog.2, Novus Biologicals™

$307.00 - $406.00

Specifications

Antigen Nanog
Clone hNanog.2
Concentration 0.2mg/mL
Immunogen Human Nanog
Gene ID (Entrez) 79923
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14330401

 
novus biologicals canada
NBP14330401MG
0.1mg Each for $406.00

NBP14330403

 
novus biologicals canada
NBP1433040025MG
0.025mg Each for $307.00
Description & Specifications

Specifications

Antigen Nanog
Clone hNanog.2
Concentration 0.2mg/mL
Immunogen Human Nanog
Gene ID (Entrez) 79923
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Western Blotting