Please login to your online account to display your discounted pricing

Mouse anti-Nanog, Clone: hNanog.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433040025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14330403

  • / Each
 

Description & Specifications

Specifications

Antigen Nanog
Applications Western Blot
Clone hNanog.2
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Murine
Immunogen Human Nanog
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 79923
Target Species Human