Please login to your online account to display your discounted pricing

Mouse anti-Nanog, Clone: hNanog.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14330401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14330401

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Western Blotting
Immunogen Human Nanog
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 79923
Clone hNanog.2
Antigen Nanog