Please login to your online account to display your discounted pricing

Mouse anti-N-Cadherin, Clone: 8C11, PE, Novus Biologicals™

$171.00 - $354.00

Specifications

Antigen N-Cadherin
Clone 8C11
Immunogen Bacterial fusion protein corresponding to the extracellular domain of N-cadherin between EC3 and EC4 amino acids 92-593.
Gene ID (Entrez) 1000
Formulation Protein G purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200231

 
novus biologicals canada
NBP200231100TESTS
100 Tests Each for $354.00

NBP20023125

 
novus biologicals canada
NBP20023125TESTS
25 Tests Each for $171.00
Description & Specifications

Specifications

Antigen N-Cadherin
Clone 8C11
Immunogen Bacterial fusion protein corresponding to the extracellular domain of N-cadherin between EC3 and EC4 amino acids 92-593.
Gene ID (Entrez) 1000
Formulation Protein G purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate PE
Format Purified
Purification Method Purified
Applications Flow Cytometry