Please login to your online account to display your discounted pricing

Mouse anti-N-Cadherin, Clone: 8C11, Biotin, Novus Biologicals™

$725.00 - $725.00

Specifications

Antigen N-Cadherin
Clone 8C11
Concentration 0.5mg/mL
Immunogen Bacterial fusion protein corresponding to the extracellular domain of N-cadherin between EC3 and EC4 amino acids 92-593.
Gene ID (Entrez) 1000
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20023255

 
novus biologicals canada
NBP200232500UG
500μg Each for $725.00
Description & Specifications

Specifications

Antigen N-Cadherin
Clone 8C11
Concentration 0.5mg/mL
Immunogen Bacterial fusion protein corresponding to the extracellular domain of N-cadherin between EC3 and EC4 amino acids 92-593.
Gene ID (Entrez) 1000
Formulation aqueous buffer, may contain carrier protein/stabilizer. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry