Please login to your online account to display your discounted pricing

Mouse anti-Myogenin, Clone: SPM144, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474002

  • / Each
 

Description & Specifications

Specifications

Antigen Myogenin
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human myogenin recombinant protein
Isotype IgG1, κ
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 4656
Reconstitution Protein A purified
Conjugate Unlabeled
Gene Accession No. P15173
Purification Method Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone SPM144
Concentration 1mg/mL
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat