Please login to your online account to display your discounted pricing

Mouse anti-Myogenin, Clone: SPM144, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Immunogen Human myogenin recombinant protein
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen Myogenin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P15173
Host Species Mouse
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Primary or Secondary Primary
Gene ID (Entrez) 4656
Reconstitution Protein A purified
Clone SPM144
Concentration 1mg/mL
Conjugate Unlabeled