Please login to your online account to display your discounted pricing

Mouse anti-Myogenin, Clone: MGN185 + F5D, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Myogenin
Clone MGN185 + F5D
Concentration 0.2mg/mL
Immunogen Human myogenin recombinant protein (MGN185); Rat myogenin recombinant fragment containing amino acid 30-224 (F5D)
Gene Accession No. P15173
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23427801

 
novus biologicals canada
NBP23427801ML
0.1mL Each for $139.00

NBP23427805

 
novus biologicals canada
NBP23427805ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Myogenin
Clone MGN185 + F5D
Concentration 0.2mg/mL
Immunogen Human myogenin recombinant protein (MGN185); Rat myogenin recombinant fragment containing amino acid 30-224 (F5D)
Gene Accession No. P15173
Gene ID (Entrez) 4656
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting