Please login to your online account to display your discounted pricing

Mouse anti-Myogenin, Clone: MGN185 + F5D, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427801

  • / Each
 

Description & Specifications

Specifications

Format Purified
Gene Accession No. P15173
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4656
Reconstitution Protein A purified
Concentration 0.2mg/mL
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen Myogenin
Formulation PBS with 0.05% BSA.
Immunogen Human myogenin recombinant protein (MGN185); Rat myogenin recombinant fragment containing amino acid 30-224 (F5D)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Clone MGN185 + F5D
Conjugate Unlabeled