Please login to your online account to display your discounted pricing

Mouse anti-Myogenin, Clone: MGN185 + F5D, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427805

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human myogenin recombinant protein (MGN185); Rat myogenin recombinant fragment containing amino acid 30-224 (F5D)
Monoclonal or Polyclonal Monoclonal
Antigen Myogenin
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Isotype IgG1, κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Protein A purified
Clone MGN185 + F5D
Conjugate Unlabeled
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Gene Accession No. P15173
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 4656
Concentration 0.2mg/mL