Please login to your online account to display your discounted pricing

Mouse anti-Myogenin, Clone: MGN185 + F5D, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23468401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23468401

  • / Each
 

Description & Specifications

Specifications

Immunogen Human myogenin recombinant protein (MGN185); Rat myogenin recombinant fragment containing amino acid 30-224 (F5D)
Primary or Secondary Primary
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Purified
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Clone MGN185 + F5D
Host Species Mouse
Quantity 0.1mg
Isotype IgG1, κ
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P15173
Conjugate Unlabeled
Antigen Myogenin
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Purification Method Purified
Gene ID (Entrez) 4656
Concentration 1mg/mL