Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: SPM427, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23288201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23288201

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Primary or Secondary Primary
Format Purified
Quantity 0.1mL
Gene ID (Entrez) 4654
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen MyoD
Regulatory Status RUO
Formulation PBS with 0.05% BSA.
Isotype IgG1, κ
Immunogen Recombinant mouse MyoD1 protein
Concentration 0.2mg/mL
Host Species Mouse
Purification Method Purified
Gene Accession No. P15172
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM427
Reconstitution Protein A purified
Conjugate Unlabeled