Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: SPM427, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen MyoD
Clone SPM427
Concentration 1mg/mL
Immunogen Recombinant mouse MyoD1 protein
Gene Accession No. P15172
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23477202

 
novus biologicals canada
NBP23477202MG
0.2mg Each for $527.00

NBP23477201

 
novus biologicals canada
NBP23477201MG
0.1mg Each for $384.00
Description & Specifications

Specifications

Antigen MyoD
Clone SPM427
Concentration 1mg/mL
Immunogen Recombinant mouse MyoD1 protein
Gene Accession No. P15172
Gene ID (Entrez) 4654
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting