missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: SPM427, Azide Free, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP23477202MG

 View more versions of this product

Catalog No. NBP23477202


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

MyoD
SPM427
Unlabeled
10mM PBS. Does not contain BSA. Azide Free.
Murine
IgG1, κ
0.2mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes a phosphor-protein of 45kDa, identified as MyoD1. The epitope of this MAb maps between amino acid 180-189 in the C-terminal of mouse MyoD1 protein. It does not cross react with myogenin, Myf5, or Myf6.
Chicken,Human,Murine,Rat
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
1mg/mL
Purified
P15172
Recombinant mouse MyoD1 protein
Purified
RUO
Primary
4654
Protein A purified