Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: SPM427, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23477201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23477201

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Antigen MyoD
Gene Accession No. P15172
Host Species Mouse
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Recognizes a phosphor-protein of 45kDa, identified as MyoD1. The epitope of this MAb maps between amino acid 180-189 in the C-terminal of mouse MyoD1 protein. It does not cross react with myogenin, Myf5, or Myf6.
Clone SPM427
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Recombinant mouse MyoD1 protein
Purification Method Purified
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4654
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled