Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: MYD712, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23469301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23469301

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human MyoD1 protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Quantity 0.1mg
Regulatory Status RUO
Isotype IgG1, κ
Host Species Mouse
Antigen MyoD
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Primary or Secondary Primary
Clone MYD712
Gene ID (Entrez) 4654
Conjugate Unlabeled
Reconstitution Protein A purified
Gene Accession No. P15172
Concentration 1mg/mL
Purification Method Purified