Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: 5.8A + MYD712, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427705

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant mouse MyoD1 protein (5.8A) and Recombinant human MyoD1 protein (MYD712)
Monoclonal or Polyclonal Monoclonal
Antigen MyoD
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P15172
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 4654
Reconstitution Protein A purified
Clone 5.8A + MYD712
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat