Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: 5.8A + MYD712, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23427701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23427701

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen MyoD
Gene Accession No. P15172
Immunogen Recombinant mouse MyoD1 protein (5.8A) and Recombinant human MyoD1 protein (MYD712)
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone 5.8A + MYD712
Concentration 0.2mg/mL
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 4654