Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: 5.8A + MYD712, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen MyoD
Clone 5.8A + MYD712
Concentration 1mg/mL
Immunogen Recombinant mouse MyoD1 protein (5.8A) and Recombinant human MyoD1 protein (MYD712)
Gene Accession No. P15172
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23454101

 
novus biologicals canada
NBP23454101MG
0.1mg Each for $384.00

NBP23454102

 
novus biologicals canada
NBP23454102MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen MyoD
Clone 5.8A + MYD712
Concentration 1mg/mL
Immunogen Recombinant mouse MyoD1 protein (5.8A) and Recombinant human MyoD1 protein (MYD712)
Gene Accession No. P15172
Gene ID (Entrez) 4654
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot