Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: 5.8A + MYD712, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454102

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Antigen MyoD
Gene Accession No. P15172
Primary or Secondary Primary
Host Species Mouse
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Immunogen Recombinant mouse MyoD1 protein (5.8A) and Recombinant human MyoD1 protein (MYD712)
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Reconstitution Protein A purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Clone 5.8A + MYD712
Gene ID (Entrez) 4654
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Quantity 0.2mg
Format Purified