Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: 5.8A + MYD712, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454101

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Antigen MyoD
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Concentration 1mg/mL
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 4654
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Gene Accession No. P15172
Isotype IgG1, κ
Clone 5.8A + MYD712
Reconstitution Protein A purified
Immunogen Recombinant mouse MyoD1 protein (5.8A) and Recombinant human MyoD1 protein (MYD712)
Purification Method Purified