Please login to your online account to display your discounted pricing

Mouse anti-MyoD, Clone: 5.8A, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454001

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Gene Accession No. P15172
Gene ID (Entrez) 4654
Primary or Secondary Primary
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.1mg
Antigen MyoD
Format Purified
Immunogen Recombinant mouse MyoD1 protein
Conjugate Unlabeled
Clone 5.8A
Regulatory Status RUO
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Reconstitution Protein A purified