Please login to your online account to display your discounted pricing

Mouse anti-Myeloid Cell Marker, Clone: SPM298, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23288110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23288110

  • / Each
 

Description & Specifications

Specifications

Antigen Myeloid Cell Marker
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM298
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Human peripheral blood mononuclear cells
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified