Please login to your online account to display your discounted pricing

Mouse anti-Myeloid Cell Marker, Clone: SPM298, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23288101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23288101

  • / Each
 

Description & Specifications

Specifications

Antigen Myeloid Cell Marker
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Isotype IgG1, κ
Quantity 0.1mL
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM298
Format Purified
Host Species Mouse
Immunogen Human peripheral blood mononuclear cells
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled