Please login to your online account to display your discounted pricing

Mouse anti-Myeloid Cell Marker, Clone: SPM298, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Myeloid Cell Marker
Clone SPM298
Concentration 1mg/mL
Immunogen Human peripheral blood mononuclear cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23475901

 
novus biologicals canada
NBP23475901MG
0.1mg Each for $384.00

NBP23475902

 
novus biologicals canada
NBP23475902MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Myeloid Cell Marker
Clone SPM298
Concentration 1mg/mL
Immunogen Human peripheral blood mononuclear cells
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)