Please login to your online account to display your discounted pricing

Mouse anti-Myeloid Cell Marker, Clone: SPM298, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23475902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23475902

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity Human
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Format Purified
Antigen Myeloid Cell Marker
Quantity 0.2mg
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Clone SPM298
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Recognizes 183kDa protein with DNA-binding characteristics, which is identified as a myeloid specific antigen.
Concentration 1mg/mL
Purification Method Purified
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Immunogen Human peripheral blood mononuclear cells
Primary or Secondary Primary
Regulatory Status RUO