Please login to your online account to display your discounted pricing

Mouse anti-MUL1, Clone: 3G2.2F7, Novus Biologicals™

$109.00 - $259.02

Specifications

Antigen MUL1
Clone 3G2.2F7
Concentration 1.0mg/mL
Immunogen Partial recombinant protein made to a C-terminal portion of the human MUL1 protein (between residues 200-352) [UniProt Q969V5]
Gene Accession No. Q969V5
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23136201

 
novus biologicals canada
NBP23136201MG
0.1mg Each for $259.02

NBP23136225

 
novus biologicals canada
NBP2313620025MG
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen MUL1
Clone 3G2.2F7
Concentration 1.0mg/mL
Immunogen Partial recombinant protein made to a C-terminal portion of the human MUL1 protein (between residues 200-352) [UniProt Q969V5]
Gene Accession No. Q969V5
Gene ID (Entrez) 79594
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting