Please login to your online account to display your discounted pricing

Mouse anti-Mucin 5AC, Clone: SPM297, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Mucin 5AC
Clone SPM297
Concentration 0.2mg/mL
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Gene Accession No. P98088
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23287901

 
novus biologicals canada
NBP23287901ML
0.1mL Each for $139.00

NBP23287910

 
novus biologicals canada
NBP23287910ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Mucin 5AC
Clone SPM297
Concentration 0.2mg/mL
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Gene Accession No. P98088
Gene ID (Entrez) 4586
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting