Please login to your online account to display your discounted pricing

Mouse anti-Mucin 5AC, Clone: SPM297, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23287901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23287901

  • / Each
 

Description & Specifications

Specifications

Antigen Mucin 5AC
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM297
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P98088
Host Species Mouse
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4586
Test Specificity Does not react to Bovine.
Reconstitution Protein A purified